Double spatulas, micro
130 mm, 40 mm, 4 mm
目录编号 BOCH3300
供应商: BOCHEM
Jump to:
Product Details & Documents
Stainless steel 18/10, anti-magnetic. With round handle, one flat and one scoop shaped, rounded spatula end.
- Straight shape
- Excellent thermal resistance, autoclavable
- Good chemical resistance
规格
- 长度:130 mm
- 刀片宽度:4 mm
- 刀片长度:40 mm