Spoon spatulas
210 mm, 12×5 mm
Catalog # BOCH3212
Supplier: BOCHEM
Jump to:
Product Details & Documents
Stainless steel. Universal spoon spatula designed for a wide range of uses.
- One end is shaped like a spoon and the other end like a spatula
- With rounded handle
- High thermal resistance, autoclavable
- Excellent resistance to chemicals
- Quick and easy to clean
Documents
Certificates
Search by Lot NumberSpecifications
- 长度:210 mm
- 线圈尺寸:12×5 mm