Forceps
Blunt, 130 mm
目录编号 BOCH1801
供应商: BOCHEM
Jump to:
Product Details & Documents
Stainless steel, PTFE coating.
- Anti-magnetic
- Very high chemical and thermal resistance
- Smooth surface prevents any contamination and makes cleaning easier
规格
- Type:Blunt
- 长度:130 mm