跳转到 页眉
跳转到 主要内容
跳转到 页脚
高级搜索
SDS
Certificates
制造商数据
0
¥0.00
订单输入
登录
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
您的账户
Cart
China
Order Entry
Order Status
您的账户
Cart
Orders Link
ContactUsLinkComponent
Home
夹具
油管夹
Hoffmann pinch cocks
Hoffmann pinch cocks
Print
Hoffmann pinch cocks
06, 38 mm
目录编号 BOCH12535
供应商:
BOCHEM
Hoffmann pinch cocks
目录编号 BOCH12535
供应商:
BOCHEM
展开图片
Return to Top
Hoffmann pinch cocks
06, 38 mm
规格
Add to cart
Jump to:
规格
Price
询价
Log in
查看库存
规格
1 * 1 件
-
+
询价
Other Product Family Options Available
Return Policy
Product Details & Documents
Nickel-plated brass, with flap.
Documents
数据表
BOCH12533_TDS_EN
规格
顺序:
06
带宽:
38 mm