Orders LinkContactUsLinkComponent
Manual microorganism identification reagents and medium, API®
Manual microorganism identification reagents and medium, API®
API® 50 CH for identification of Bacillus, 10 strips, 10 Vial
目录编号 BMER50300
Marketsource
供应商:   BIOMERIEUX
Manual microorganism identification reagents and medium, API®
目录编号 BMER50300
供应商:  BIOMERIEUX
Marketsource
CAS号:  
Manual microorganism identification reagents and medium, API®
API® 50 CH for identification of Bacillus, 10 strips, 10 Vial
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 10 Vial
Product Details & Documents
API® Identification kits provide simple methodology for detection of various groups of organisms and also enzymes. It includes the evolution of international taxonomy, description of the new bacterial species and newly acquired bacteriology data.
  • Rapid and reliable method for bacterial identification
Documents
Certificates
按批号搜索
规格
规格
  • 包装规格:
    10 Vial
  • 说明:
    API® 50 CH for identification of Bacillus, 10 strips
规格