十二烷基硫酸钠 85%, pure
3401, Plastic bottle, 100 g
Jump to:
Product Details & Documents
同义词: SDS, 月桂基硫酸盐, 月桂基硫酸钠
- CAS号:151-21-3
- 化学式:CH₃(CH₂)₁₁OSO₃Na
- MDL 编号:MFCD00036175
- Molecular Weight:288.38 g/mol
- 默克索引:14,08636
- 熔点:206 °C
- 密度:1,1 g/cm³ (20 °C)
- 闪点:>150 °C
- 储存温度:Ambient
- EINECS:205-788-1
- ADR:4.1,III
Documents
ACRO22614
SDS
规格
规格测试结果
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- GC:≥80 %
规格
- 包装类型:Plastic bottle
- 包装规格:100 g
- 顺序:3401



