Accessories for analytical Mill, A 11 basic
02, 250 ml grinding chamber, Use with double beater 431-2113. Polycarbonate with stainless steel inlet. Not suitable for Use with liquid nitrogen
目录编号 IKAA2904100
供应商: IKA JANKE KUNKEL
Jump to:
Product Details & Documents
规格
- 说明:250 ml grinding chamber
- 顺序:02
- 用于:Use with double beater 431-2113. Polycarbonate with stainless steel inlet. Not suitable for Use with liquid nitrogen