跳转到 页眉
跳转到 主要内容
跳转到 页脚
高级搜索
SDS
Certificates
制造商数据
0
¥0.00
订单输入
登录
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
您的账户
Cart
China
Order Entry
Order Status
您的账户
Cart
Orders Link
ContactUsLinkComponent
Home
烧瓶
容量瓶
Volumetric flasks, class B
Volumetric flasks, class B
Print
Volumetric flasks, class B
250 ml, 19/26, 235 mm
目录编号 HIRS7560186
供应商:
HIRSCHMANN
Volumetric flasks, class B
目录编号 HIRS7560186
供应商:
HIRSCHMANN
展开图片
Return to Top
Volumetric flasks, class B
250 ml, 19/26, 235 mm
规格
Add to cart
Jump to:
规格
Price
询价
Log in
查看库存
规格
1 * 2 件
-
+
询价
Other Product Family Options Available
Return Policy
Product Details & Documents
PMP, translucent, with PP NS-stopper.
规格
模拟器:
19/26
容量:
250 ml
高度:
235 mm