Automatic burettes, Pellet pattern, lateral stopcock
25 ml, 0,05 ml, 500 mm, 0,045 ± ml
目录编号 HIRS341037060
供应商: HIRSCHMANN
Jump to:
Product Details & Documents
Automatic burettes, Pellet pattern, class B, DURAN® with blue graduation.
- Schellbach stripes for enhanced legibility
- Lateral glass stopcock or PTFE valve stopcock
Certifications: Caliberated for DIN EN ISO 385.
Documents
HIRS341016060_TDS_EN
Datasheet
规格
- 容量:25 ml
- 分度:0,05 ml
- 误差:0,045 ± ml
- 刻度长度:500 mm