Milliflex® Quantum Camera
Quantum camera, for use with Milliflex® Quantum systems
目录编号 MILXMXQCAM001
New Product
供应商: MILLIPORE
Jump to:
Product Details & Documents
The Milliflex® Quantum camera connected to a PC makes sample reading more convenient during the Milliflex® filtration test of filtrable samples, including in-process and final product samples (pharmaceuticals and water monitoring).
Material: 304L stainless steel plate (front) aluminium housing, epoxy-coated housing glass window(s)
Application: Pharmaceutical
Shipped in: Ambient
Input and output: Powered by the computer used
The Milliflex® Quantum camera combined with the Milliflex® Quantum Spot Counter software makes the sample reading more convenient and allows tracking of records for optimised microbiology QC labs.
规格
- 说明:Quantum camera, for use with Milliflex® Quantum systems



