Masterflex® L/S Digital Drive Systems
Supplier: MASTERFLEX PROPRIETARY
Certificates
About this item
Don't lose control of your metering applications - digital display gives you repeatable control.
- Flow rate 0,06 to 3400 ml/min - flow rate depends on drive rpm, pump head, and tubing size
- Three digit LED display shows motor rpm - confirm speed setting at a glance
- Reversible motor - purge tubing before or after pumping; pump fluid in either direction
- LED indicators confirm motor direction
- Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
- Separate speed and power (on/off) control - maintain speed setting while turning pump on/off
- Stackable, splash-resistant ABS plastic housing; IP 33 rated
- Remote control capabilities via a DB9 female connector on back of drive
- Speed control input: 4 to 20 mA or 0 to 10 VDC
- Pump direction (CW/CCW) via open collector or contact closure
- Start/stop via open collector or contact closure
- Membrane keypad for easy up-front control of speed, direction, and start/stop
- Automatic tubing retention
- Fixed occlusion
These accurate analogue console drives have powerful motors, speed display, and ±0,25% speed control for precise flow regulation - ideal for metering applications. Remote speed control requires DB9 male connector 07595-45 or cable assembly 07595-47. Use remote connector for control of start/stop via contact closure or open collector (5 V TTL); control start/stop via footswitch 77595-35 or handheld remote 07528-80. This system includes the drive and pump head.
Accessories information:
Accessories Information: Drive 07528-10 and pump head 77200-62.
Specifications
- Catalog No:
- MFLA77712-37
- MFLA77712-36
- 型号:
- Masterflex L/S Drive with Easy-Load II Pump Head
- Masterflex L/S Drive with Easy-Load II Pump Head
- 通道:
- 1
- 1
- 滚筒数量:
- 4; stainless steel
- 4; stainless steel
- 外壳材质:
- Polyphenylene Sulfide (PPS)
- Polyphenylene Sulfide (PPS)
- 输入:
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure (remote inputs)
- 4 - 20 mA, 0 - 10 V, start/stop and direction via contact closure (remote inputs)
- 功率:
- 90 - 260 VAC, 50/60 Hz
- 90 - 260 VAC, 50/60 Hz
- 管尺寸:
- L/S® 15, L/S 24, L/S 35, L/S 36
- L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
- 质保:
- 2 years
- 2 years