Hydrophobic interaction chromatography columns, TSK-GEL®
Catalog # TOYO0007573
Supplier: TOSOH BIOSCIENCE
Specifications
- 列格式:Analytical
- 内径:7,50 mm
- 颗粒大小:10 µm
- 孔大小:1000 Å
- 说明:Phenyl-5PW
- 长度:75 mm
- Phase:Phenyl-5PW
- 包装规格:1
Specifications
About this item
These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.
- Rigid polymeric base resin
- Eliminates undesirable interactions with column hardware
- Wide pH range (2 to 12) enables robust cleaning options