二苄基二硫 ≥98.0% (by GC)
3308, Glass bottle, 25 g
Catalog # TCIAB0417-25G
Supplier: TCI
Jump to:
Product Details & Documents
Synonyms: 二硫化苄
- CAS Number:150-60-7
- Formula:C₁₄H₁₄S₂
- MDL Number:MFCD00004783
- Molecular Weight:246.4 g/mol
- Merck Index:14,03013
- Boiling Pt:142 - 148 °C (0,05 -rr)
- Melting Pt:62…67 °C
- Density:1,302 g/cm³ (25 °C)
- EINECS:205-764-0
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:70.0 to 73.0 deg-C
- Solubility in hot Methanol:within almost transparency
Specifications
- 包装类型:Glass bottle
- 包装规格:25 g
- 顺序:3308
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



