Measuring cylinders, tall form, class A, borosilicate glass, with DKD certificate
Catalog # HIRS2240180DKD
Supplier: HIRSCHMANN
Specifications
- Type:with DKD certificate
- 容量:100 ml
- 高度:260 mm
- 分度:1,0 ml
- 包装规格:1
- 误差:0,5 ± ml
Specifications
About this item
DURAN® Borosilicate glass, Class A.
- Blue graduation, main point ring graduation
- Hexagonal glass base, spout
- With dated batch identification
With DAkkS-Certificate.