Orders LinkContactUsLinkComponent
WHEATON® Sample Vials in Lab File®
WHEATON® Sample Vials in Lab File®
供应商:   DWK LIFE SCIENCES
WHEATON® Sample Vials in Lab File®
供应商:  DWK LIFE SCIENCES
Certificates
WHEATON® Sample Vials in Lab File®
WHEATON® Sample Vials in Lab File®
5 Options Available Below
Product Details & Documents
Clear vials made from low extractable borosilicate glass, amber vials made from borosilicate glass.
  • Sample vials with caps attached to vial help maintain cleanliness
  • Lab File® with partitioned trays provides an easy way to inventory samples or to store empty vials
  • Clear vials conform to ASTM E438 Type I, Class A and USP Type I requirements
  • Amber vials conform to USP Type I requirements for light transmission to protect light-sensitive products
  • 14B rubber lined black phenolic screw cap for aqueous samples
Product Family Options
Standard, clear
产品信息
  • 顺序
    说明
    容量
    直径×高度
    颜色
    Cap included
    Cap type
    盖颜色
    隔膜
    可用库存
    Price
  • 09
    Sample vials in Lab File™ with caps attached
    40,00 ml
    28×98 mm
    透明
    Solid top
    Colour-21390420|Colour-21390392
    PTFE/14B rubber
    Log in 查看库存
    Log in 查看库存
    1 * 72 件
    1 * 72 件
Standard, amber
产品信息
  • 顺序
    说明
    容量
    直径×高度
    颜色
    Cap included
    Cap type
    盖颜色
    隔膜
    可用库存
    Price
  • 11
    Sample vials in Lab File™ with caps attached
    4,00 ml
    15×48 mm
    琥珀色
    Solid top
    Colour-21390420|Colour-21390392
    PTFE/14B rubber
    Log in 查看库存
    Log in 查看库存
    1 * 144 件
    1 * 144 件
  • 12
    Sample vials in Lab File™ with caps attached
    4,00 ml
    15×48 mm
    琥珀色
    Solid top
    Colour-21390420|Colour-21390392
    PTFE/14B rubber
    Log in 查看库存
    Log in 查看库存
    1 * 144 件
    1 * 144 件
  • 15
    Sample vials in Lab File™ with caps attached
    20,00 ml
    28×60 mm
    琥珀色
    Solid top
    Colour-21390420|Colour-21390392
    PTFE/14B rubber
    Log in 查看库存
    Log in 查看库存
    1 * 72 件
    1 * 72 件
  • 16
    Sample vials in Lab File™ with caps attached
    40,00 ml
    28×95 mm
    琥珀色
    Solid top
    Colour-21390420|Colour-21390392
    PTFE/14B rubber
    Log in 查看库存
    Log in 查看库存
    1 * 72 件
    1 * 72 件