Capillary columns, Elite-1 for autosystem
Catalog # PERCN9316051
Supplier: PERKIN ELMER GC INSTRUMENTS
Specifications
- 列格式:Capillary
- 薄膜厚度:3,00 µm
- 内径:0,53 mm
- 长度:30 m
- Phase:Elite-1
- 包装规格:1
Specifications
About this item
The Elite-1 columns are 100% dimethyl polysiloxane and come in a variety of sizes.