Orders LinkContactUsLinkComponent
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 mL
目录编号 PROMV4231
供应商:   PROMEGA
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
目录编号 PROMV4231
供应商:  PROMEGA
CAS号:  
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
Buffer, EDTA buffer solution pH 8,0 (0,5 mol/L)
EDTA buffer solution pH 8,0 (0,5 mol/l), Glass bottle, 100 mL
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 100 毫升
Product Details & Documents
This EDTA solution is a chelator of divalent cations and is suitable for biochemistry and molecular biology applications
  • Available in 0,5 M or 10 mM concentrations
  • 10 mM concentration also provided as part of Maxwell® HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)
It is supplied as a solution in deionized water.
规格
规格
  • 包装类型:
    Glass bottle
  • 包装规格:
    100 mL
  • 说明:
    EDTA buffer solution pH 8,0 (0,5 mol/l)
规格