Test tube racks, Stack Rack™, SP Bel-Art
BELAF188601316
- 用于管径:13 - 16 mm
- 孔数量:40 (4×10)
- 材质:PP
- 顺序:02
- 宽度×深度×高度:270×112×133 mm
- 包装规格:1
PP, white
- Sturdy and chemical resistant
- Will not shatter or stain
- With top shelf height of 70 mm
These rigid racks features interlocking end plates with handles for secure stacking when loaded with tubes up to 100 mm long. Rows are numbered and lettered for easy tube identification.