Racks, two tier, for microcentrifuge tubes, SP Bel-Art
Catalog # BELAF188430000
Supplier: Bel-Art Products, a Part of SP
Specifications
- 用于管:1,5 ml
- 孔数量:20 (2×10)
- 宽度×深度×高度:210×70×35 mm
- 包装规格:1
Specifications
About this item
This two tier PP rack holds microcentrifuge tubes in numbered wells and tubes are held roughly protruding above the rack surface for easy handling.