二氯甲烷 ≥99.9% (by GC), ChromAR® for HPLC, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # MACR4879-10
Supplier: MACRON AVANTOR BRAND
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- 包装类型:Glass bottle, amber, narrow mouth
- 包装规格:4 L
- 顺序:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:≥99.9%
- ACS - Assay (CH₂Cl₂) (by GC, corrected for water):≥99.9 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.0003 %
- ACS - Titrable Acid (meq/g):≤0.0003
- ACS - Free Halogens:Passes Test
- ACS - Water (H₂O)(by coulometry):≤0.01 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 235 nm:≤1.0
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm:≤0.12
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 260 nm:≤0.04
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 - 400 nm:≤0.01
- Liquid Chromatography Suitability Absorbance:Passes Test
Specification Test Results
About this item
:
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:39,8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1,32 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
:GHS08
:GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881