Printer, impact, CBM910
901, Serial printer, CBM910
目录编号 BELL55-14
供应商: BELLINGHAM STANLEY
Product Details & Documents
Dot matrix, miniature, 24 column, RS232, impact printer that can be used to provide hard copy of refractometer or polarimeter measurement data for FDA regulated environments or GLP audit trail as dot matrix impact printers are the preferred option for data longevity compared to thermal type printers.
- Dot matrix for longevity of print
- 24 column width
- Small footprint
Bellingham + Stanley RFM refractometers and ADP polarimeters print outputs are optimised to print with this 24-column printer, making this a convenient printer for all models.
Delivery information: Supplied with replaceable black ink ribbon.
规格
- 说明:Serial printer, CBM910
- 顺序:901
Accessories
产品信息
- 目录编号顺序说明用于可用库存Price