Orders LinkContactUsLinkComponent
记忆型温湿计,Traceable®
记忆型温湿计,Traceable®
供应商:   VWR Collection
记忆型温湿计,Traceable®
供应商:  VWR Collection
Certificates
记忆型温湿计,Traceable®
记忆型温湿计,Traceable®
2 Options Available Below
Product Details & Documents
该款温湿计可测量温度和相对湿度,并将结果显示在双LCD显示屏上。装置配有输出插座,用于将数据传输至电脑。每次按键输入都会听到按键音,便于确认输入。适用于监测洁净室、计算机室、存储设施和任何地方会影响重要组件和试验的温度和相对湿度。
  • Maintains accuracy with 'do it yourself' calibration
  • Simultaneously displays both temperature and relative humidity on dual-display LCD (28,6 mm character height)
  • Minimum/maximum memory
  • Interface for transfer of data to a computer
  • An audible click confirms each key entry
Compliant ISO/IEC 17025, ANSI/NSCL Z540-1 and 9001.
Certifications: Compliant ISO/IEC 17025, ANSI/NSCL Z540-1 and 9001. Traceable to NIST standards.
Delivery information: Supplied with probe (Lר: 216×17 mm) and 91 cm cable, Traceable® certificate of calibration, hard plastic carrying case and 9 volt battery for 100 hours of continuous use. The data acquisition software incl. cable (620-2048) that needs to be ordered separately.
Caution: *mid range
Product Family Options
产品信息
  • 顺序
    说明
    范围
    准确度
    分辨率
    宽度×深度×高度
    重量
    可用库存
    Price
  • 01
    Memory hygrometer/thermometer
    −18…+93 °C; 1 - 95% RH
    ±1.0 (0…40); 2.0 °C; ±2.0% RH(中间范围); ±4.0% RH
    0,1 °C; 0,1% RH
    181×32×70 mm
    305 g
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥6,549.00
    ¥6,549.00
    1 * 1 件
       ¥6,549.00
  • 01
    Memory hygrometer/thermometer with dew
    point: 0 to 50 °C
    −18…+93 °C; 10 - 95% RH
    ±1,0 (0…40); 2,0 °C; ±2,0% RH*; ±4,0% RH
    0,1 °C; 0,1% RH
    181×32×70 mm
    305 g
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件 - ¥13,967.00
    ¥13,967.00
    1 * 1 件
       ¥13,967.00