Orders LinkContactUsLinkComponent
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Dissection boards, DissecTable™
Supplier:  SIMPORT SCIENTIFIC
Certificates
Dissection boards, DissecTable™
Dissection boards, DissecTable™
3 Options Available Below
Product Details & Documents
HDPE / PS. These dissection boards feature a large cutting area on one side. They include dimensional scales in inches and centimeters, a grid and dimensional circles of different sizes. The other side offers two cutting boards half the size with the same dimensional features printed on each one of them.
  • Drain groove is carved all around the edge of the boards in order to contain fluids
  • All corners have rubber feet giving more stability to the working surface
  • Bases are stackable and will not move sideways during dissecting work
Product Family Options
Cutting boards
Product Information
  • 说明
    颜色
    宽度×深度×高度
    Availability
    Price
  • Small board with 3 cutting areas
    白色
    330,2×279,4×12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
  • Large board with 3 cutting areas
    白色
    575×400×12,5 mm
    Log in for availability
    Log in for availability
    1 * 1 件
    1 * 1 件
Product Information
  • 说明
    颜色
    宽度×深度×高度
    Availability
    Price
  • Board base
    蓝色
    481×656×91 mm