Orders LinkContactUsLinkComponent
PCR和存储用铝箔密封(96孔板)
PCR和存储用铝箔密封(96孔板)
供应商:  VWR Collection
Certificates
PCR和存储用铝箔密封(96孔板)
PCR和存储用铝箔密封(96孔板)
3 Options Available Below
Product Details & Documents
Aluminium foils, 75,7 µm thick, for use with 96 or 384 well plates. Fit inside the rim of raised rim plates. These foils have one partial width end tab. Available non sterile only.
  • 建议用于−80到+120 °C
  • 温度,无DNase、无RNase及核酸认证
每个箔的长×深:127.0×77.8 mm,包含单个9.5 mm端部凸片。
Product Family Options
产品信息
  • 顺序
    说明
    Optical property
    厚度
    灭菌
    可用库存
    Price
  • 01
    96孔板用铝箔
    Light blocking
    75,7 µm
    -
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件 - ¥1,586.00
    ¥1,586.00
    1 * 100 件
       ¥1,586.00
  • 02
    384孔板用铝箔
    Light blocking
    75,7 µm
    -
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件 - ¥2,192.00
    ¥2,192.00
    1 * 100 件
       ¥2,192.00
  • 02
    Aluminium foils for 384-well plates
    Light blocking
    75,7 µm
    -
    Log in 查看库存
    Log in 查看库存
    1 * 100 件
    1 * 100 件 - ¥2,106.00
    ¥2,106.00
    1 * 100 件
       ¥2,106.00