Stirring spatulas
01, Standard, 120 mm, 3 mm, 透明
目录编号 BOET09.241.0100
供应商: BOETTGER PAUL
Jump to:
Product Details & Documents
PS, transparent. Stirring rod for a wide variety of laboratory applications.
- With or without mixing plate at one end
These spatulas can be used for various applications, such as mixing samples and solutions in test tubes, adding small reagent volumes and releasing blood clots from the side of test tube walls.
规格
- 颜色:透明
- 说明:Standard
- 长度:120 mm
- 顺序:01
- 刀片宽度:3 mm