Orders LinkContactUsLinkComponent
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
04, Decapper for 20 mm crimp caps
目录编号 WHEAW225353
供应商:   DWK LIFE SCIENCES
Decappers, E-Z Decappers™, WHEATON®
目录编号 WHEAW225353
供应商:  DWK LIFE SCIENCES
Decappers, E-Z Decappers™, WHEATON®
Decappers, E-Z Decappers™, WHEATON®
04, Decapper for 20 mm crimp caps
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
These decappers are used to remove aluminium seals from bottles and vials with crimp or serum finish.
  • Cushioned ergonomic handle reduces hand fatigue
  • Labeled for quick size identification
  • Autoclavable
规格
  • 说明:
    Decapper for 20 mm crimp caps
  • 顺序:
    04