Arsenic standard solution, 1000 mg/L As in 0,5 M nitric acid (from H₃AsO₄), Certipur® standard for AAS, Supelco®
目录编号 MERC1.19773.0500
供应商: MERCK MILLIPORE
CAS 编号:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
规格
- 浓度:1000 mg/L
- 基质:0,5 M HNO₃
- 包装类型:Plastic bottle
- 包装规格:500 mL
- M-Clarity quality level:MQ100
- 元件:砷
- 符号:As
规格
关于此项目
Certifications: Traceable to SRM from NIST.