Trypsinizing flasks, WHEATON®
150 ml, Top screw cap 38-430, Screw cap 20-415
Catalog # WHEA355804
Supplier: DWK LIFE SCIENCES
Jump to:
Product Details & Documents
Borosilicate glass, clear.
- With pour out and caps
- Fluted design improves mixing
- Deep skirted screw caps
These flasks feature deep-fluted sides to promote mixing. A dam below the sidearm retains untrypsinized tissue when decanting. A deep skirted screw cap on the sidearm prevents contamination of flask contents. The sidearm allows easier decanting of contents while retaining large particles behind the dam. Sidearm caps are 20-415 size. Caps have white styrene-butadiene rubber liners.
规格
- 容量:150 ml
- 说明:Top screw cap 38-430
- 管颈:Screw cap 20-415