Counting chambers
Slides with ten chambers and patented counting grids (table included), with grid, 3×3 mm
供应商: KOVA INTERNATIONAL
Jump to:
Product Details & Documents
Designed to reduces pipetting errors and eliminate variable that could interfere with microscopic urinalysis (UA).
- High optical quality through the composition acrylate
- Perimeter contained wells for precise volume control and protection of cross-contamination and leakage
- Capillary action for reproducible precision
- Homogeneous suspension of sample sediments: Retained volume of 1 µl
- For grid models, the grid is made up of nine squares, each divided into nine smaller squares
Grid: 3×3 mm
Volume retained on the grid: 1 µl
Documents
规格
- Type:with grid
- 宽度×深度:3×3 mm
- 说明:Slides with ten chambers and patented counting grids (table included)