Orders LinkContactUsLinkComponent
Test strips, phosphate, MQuant®, Supelco®
Test strips, phosphate, MQuant®, Supelco®
Phosphate test, 10-25-50-100-250-500 mg/l PO₄, Phosphormolybdenum blue, MQ, 100 Tests
目录编号 MERC1.10428.0001
供应商:   MERCK MILLIPORE
Test strips, phosphate, MQuant®, Supelco®
目录编号 MERC1.10428.0001
供应商:  MERCK MILLIPORE
CAS号:  
Test strips, phosphate, MQuant®, Supelco®
Test strips, phosphate, MQuant®, Supelco®
Phosphate test, 10-25-50-100-250-500 mg/l PO₄, Phosphormolybdenum blue, MQ, 100 Tests
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 100 Tests
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
  • Easy to handle: Just dip, add reagent, wait and measure visually against colour scale
  • Immediate results within a few minutes enables fast decisions
  • Portable: Can be used inside or outside the laboratory
Documents
规格
规格
  • 化学方法:
    Phosphormolybdenum blue
  • 包装规格:
    100 Tests
  • 说明:
    Phosphate test
  • 类型:
    MQ
  • 范围:
    10-25-50-100-250-500 mg/l PO₄
  • Cat. No.:
    MERC1.10428.0001
  • M-Clarity quality level:
    MQ100