过氧化钠 ACS
3401, Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: 超氧化钠
- CAS Number:1313-60-6
- Formula:Na₂O₂
- MDL Number:MFCD00003497
- Molecular Weight:77.98 g/mol
- Merck Index:13,08728
- Melting Pt:460 °C (dec.)
- Density:2,8 g/cm³
- Storage Temperature:Ambient
- EINECS:215-209-4
- UN:1504
- ADR:5.1,I
Documents
ACRO31576
SDS
Specifications
Specification Test Results
- Appearance:Yellow Powder or granules (May contain a small quantity of discolored material from the manufacturing process)
- Titration with KMnO4:≥93.0 %
- Chloride (Cl):≤20 ppm
- Phosphate (PO4):≤5 ppm
- Sulfate (SO4):≤10 ppm
- Iron (Fe):≤50 ppm
- Heavy metals (as Pb):≤20 ppm
Specifications
- 包装类型:Plastic bottle
- 包装规格:100 g
- 顺序:3401