水, Ultrapure for CE
2725, 500 mL
目录编号 HEWL5062-8578
Jump to:
Product Details & Documents
Premade buffers help eliminate the time-consuming buffer preparation process. All Agilent buffers and reagents are designed to meet the stringent demands of CE.;To maintain high reproducibility, all buffer solutions are standardized to 0.02 pH units under "class 10" clean room conditions. Superior quality control ensures reproducible results bottle-to-bottle and batch-to-batch.
- Manufactured under GLP/GMP conditions in ISO9001 facilities
- Chemicals are all electrophoresis grade, with nearly all ionic and organic impurities removed
- Solutions are prefiltered through 0.2 mm filters to ensure removal of particulates
- All buffer solutions are standardized to 0.02 pH units in accredited laboratories
- Each buffer shipped with assay information and verification of purity
同义词: Hydrogen oxide
- CAS号:7732-18-5
- 化学式:H₂O
- MDL 编号:MFCD00011332
- Molecular Weight:18.02 g/mol
- 默克索引:12,10175
- 沸点:100 °C (1013 hPa)
- 熔点:0 °C
- 密度:1 g/cm³ (4 °C)
- 储存温度:Ambient
- EINECS:231-791-2
规格
规格
- 包装规格:500 mL
- 顺序:2725



