Orders LinkContactUsLinkComponent
Brushes for flasks
Brushes for flasks
供应商:   BOCHEM
Brushes for flasks
供应商:  BOCHEM
Certificates
Brushes for flasks
Brushes for flasks
4 Options Available Below
Product Details & Documents
Brushes for flasks, iron wire, zinced with natural bristles.
Product Family Options
产品信息
  • 顺序
    Bristle colour
    Brush head length
    Brush head Ø
    Handle material
    总长度
    Handle length
    可用库存
    Price
  • 01
    自然色
    130 mm
    50 mm
    380 mm
    250 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 02
    自然色
    140 mm
    55 mm
    420 mm
    280 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 03
    自然色
    120 mm
    63 mm
    480 mm
    360 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 04
    自然色
    190 mm
    80 mm
    550 mm
    360 mm