Orders LinkContactUsLinkComponent
Brushes for flasks
供应商:   BOCHEM
Brushes for flasks
供应商:  BOCHEM
Certificates
Brushes for flasks
Brushes for flasks
3 Options Available Below
Product Details & Documents
Brushes for flasks, iron wire, zinck plated, with natural bristles.
Product Family Options
产品信息
  • 顺序
    Bristle colour
    Brush head length
    Brush head Ø
    Handle material
    总长度
    Handle length
    可用库存
    Price
  • 01
    自然色
    120 mm
    50 mm
    340 mm
    220 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 02
    自然色
    140 mm
    55 mm
    420 mm
    280 mm
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 03
    自然色
    170 mm
    60 mm
    470 mm
    300 mm