Orders LinkContactUsLinkComponent
Volumetric flasks, class B, Azlon®
Volumetric flasks, class B, Azlon®
供应商:   DWK LIFE SCIENCES
Volumetric flasks, class B, Azlon®
供应商:  DWK LIFE SCIENCES
Certificates
Volumetric flasks, class B, Azlon®
Volumetric flasks, class B, Azlon®
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PMP (TPX®), transparent, with PE stopper.
  • Clear, easy to read calibration line
  • Exposure to temperature up to 121 °C (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to 60 °C is recommended to preserve marks and inscriptions
Error limits according to class B, DIN EN ISO 1042.
Product Family Options
产品信息
  • 容量
    模拟器
    高度
    误差
    可用库存
    Price
  • 100 ml
    14/23
    180 mm
    0,20 ± ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 50 ml
    10/19
    150 mm
    0,12 ± ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 25 ml
    10/19
    115 mm
    0,08 ± ml
    Log in 查看库存
    Log in 查看库存
    1 * 1 件
    1 * 1 件
  • 250 ml
    19/26
    235 mm
    0,30 ± ml