Pasteur pipettes
3, 3 ml, 154 mm, 0,50 ml, +, Individually wrapped
供应商: KARTELL
Jump to:
Product Details & Documents
PE, disposable, sterile or non sterile.
- Disposable PE pipettes with bulb and graduations
- The slender tip guarantees precision transfer of the sample
- Non sterile pipettes are available in multi-packs; sterile pipettes are individually wrapped
: *Capillary
规格
- 包装规格:Individually wrapped
- 灭菌:+
- 容量:3 ml
- 长度:154 mm
- 顺序:3
- 分度:0,50 ml