ContactUsLinkComponent
Condensers, Allihn, for Soxleth extractors
Condensers, Allihn, for Soxleth extractors
Catalog # LENZ05226005
Supplier:  LENZ LABORGLAS
undefined
Condensers, Allihn, for Soxleth extractors
Catalog # LENZ05226005
Supplier:  LENZ LABORGLAS
Specifications
  • 圆锥体大小:
    34/35 NS
  • 用于:
    Soxleth extractor 70 ml
  • 外套管长度:
    200 mm
  • 顺序:
    02
  • 包装规格:
    1
Specifications
About this item
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With NS cone. Water connection with glass nozzles.