Sedimentation tubes
Sedimentation tubes, 16×105 mm, 3000×g, 1000/bag, 2 bag/pack
目录编号 BRND114740
供应商: BRAND
Jump to:
Product Details & Documents
PS, transparent.
- Conical bottom
规格
- 直径×长度:16×105 mm
- 包装规格:1000/bag, 2 bag/pack
- 说明:Sedimentation tubes
- 最大相对离心力:3000×g