订单输入
China
Orders LinkContactUsLinkComponent
N-氯代酞酰亚胺 ≥95.0% (by titrimetric analysis)

N-氯代酞酰亚胺 ≥95.0% (by titrimetric analysis)

腐蚀性
供应商:   TCI
N-氯代酞酰亚胺 ≥95.0% (by titrimetric analysis)
供应商:  TCI
Certificates
N-氯代酞酰亚胺 ≥95.0% (by titrimetric analysis)
N-氯代酞酰亚胺 ≥95.0% (by titrimetric analysis)

2 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS号:3481-09-2
  • 化学式:C₈H₄ClNO₂
  • MDL 编号:MFCD00023027
  • Molecular Weight:181.58 g/mol
  • EINECS:222-459-8
  • UN:1759
  • ADR:8,II
Specifications for Product Family

规格测试结果

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 95.0 %
  • Melting point:
    186.0 to 190.0 deg-C
Product Family Options

产品信息

  • 顺序
    包装类型
    包装规格
    可用库存
    Price
  • 3308
    Glass bottle
    25 g
    Log in 查看库存
    Log in 查看库存
    1 * 25 克
    1 * 25 克

  • 3645
    Glass bottle
    500 g
    Log in 查看库存
    Log in 查看库存
    1 * 500 克
    1 * 500 克

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.