Orders LinkContactUsLinkComponent
Staining troughs, Coplin
Staining troughs, Coplin
Staining troughs acc. to Coplin, for 10 slides, with lid, 80 mm, 115 mm
目录编号 BRND472800
供应商:   BRAND
Staining troughs, Coplin
目录编号 BRND472800
供应商:  BRAND
Staining troughs, Coplin
Staining troughs, Coplin
Staining troughs acc. to Coplin, for 10 slides, with lid, 80 mm, 115 mm
Jump to:
Price
询价
Log in 查看库存
1 * 10 件
Product Details & Documents
Soda-lime glass.
For10 standard microscopy slides (76×26 mm).
规格
  • 直径:
    80 mm
  • 说明:
    Staining troughs acc. to Coplin, for 10 slides, with lid
  • 高度:
    115 mm