Wipes for delicate tasks, KIMTECH SCIENCE* KIMWIPES*
KIMB21905-026_U
- 包装规格:1 box of 280 sheets
- 颜色:白色
- 说明:KIMTECH SCIENCE* KIMWIPES* EX-L delicate task wipes, 1 ply
- L×W:213×114 mm
- 顺序:01
- Environmentally Preferable:
KIMWIPES* are suitable for general multipurpose and delicate task cleaning. The creped surface picks up and holds oil, dust, and liquids from glass, plastic, metal, and wood surfaces. The wipes are made of 100% virgin fiber, which enables cleaning without scratching or leaving a residue. Perfect for laboratory and research environments.
- Extra-low-lint, low-extractable
- Space-saving Pop-Up* dispenser box with antistatic LINTGUARD* polyshield