Cleanroom boot covers, Kimtech™ A5
Catalog # KIMB88808
Supplier: KIMBERLY CLARK
Specifications
- 滑轨尺寸:U*
- 灭菌:+
- Garment application:Cleanroom
- 颜色:白色
- 说明:Wrap-around vinyl foot
- 顺序:04
- Environmentally Preferable:Waste Reducing
- 包装规格:100 PAIR
- 洁净室等级:100/ISO 5
Specifications
About this item
The sterile KIMTECH PURE* cleanroom boot covers are made of SMS (Spunbond Meltblown Spunbond) material that gives them a cloth-like feeling. Available with standard vinyl sole or with wrap-around vinyl foot.
- Wrap-around vinyl foot is seamed above the floor line to reduce risk of liquid penetration and trip hazards Tunneled elastic and bound seams Two extra long ties
Suitable for use in Class 100, ISO 5, M3.5 environments.
*One size fits all
Certifications: PPE Cat I (EU) 2016/425
Packaging: Vacuum packed in pairs for added sterility assurance.