Orders LinkContactUsLinkComponent
洁净室实验服,VWR® Maximum,SF
洁净室实验服,VWR® Maximum,SF
供应商:   VWR Collection
洁净室实验服,VWR® Maximum,SF
供应商:  VWR Collection
Certificates
洁净室实验服,VWR® Maximum,SF
7 Options Available Below
Product Details & Documents
VWR® Maximum SF lab coats are the cleanest and most durable within the entire VWR® clothing line. Made from spunbonded polypropylene fabric (SPP) and coated with breathable polyethylene film, these garments have been designed to optimise barrier protection, cleanliness, and comfort properties.
  • Tapered collar, inset sleeves, snap closure, elasticated cuffs and three pockets
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding: Level I Helmke drum classification
  • Light chemical splash resistance
  • Improved breathability to ensure user comfort
  • Soft, cloth-like fabric with maximum tear strength
在经ISO认证的工厂进行测试和制造,并有着严格的程序控制。由独立实验室测试进行批量控制和检验。
Specifications for Product Family
规格
  • Catalog No:
  • VWRI113-1138
  • VWRI113-1139
  • VWRI113-1140
  • VWRI113-1141
  • VWRI113-1142
  • VWRI113-1143
  • VWRU414004-703
  • 关闭类型:
  • 带按扣紧固
  • 带按扣紧固
  • 带按扣紧固
  • 带按扣紧固
  • 带按扣紧固
  • 带按扣紧固
  • 带按扣紧固
  • 袋:
  • 带口袋
  • 带口袋
  • 带口袋
  • 带口袋
  • 带口袋
  • 带口袋
  • 带口袋
  • Garment application:
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
  • Cleanroom|Laboratory
规格
Product Family Options
产品信息
  • 顺序
    颜色
    衣服尺寸
    长度
    可用库存
    Price
  • 01
    白色
    S
    100 cm
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥838.00
    ¥838.00
    1 * 25 件
       ¥838.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI113-1138
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory
  • 03
    白色
    M
    100 cm
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥871.00
    ¥871.00
    1 * 25 件
       ¥871.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI113-1139
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory
  • 05
    白色
    L
    100 cm
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥916.00
    ¥916.00
    1 * 25 件
       ¥916.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI113-1140
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory
  • 07
    白色
    XL
    100 cm
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥946.00
    ¥946.00
    1 * 25 件
       ¥946.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI113-1141
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory
  • 09
    白色
    XXL
    100 cm
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥1,034.00
    ¥1,034.00
    1 * 25 件
       ¥1,034.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI113-1142
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory
  • 11
    白色
    3XL
    100 cm
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥1,045.00
    ¥1,045.00
    1 * 25 件
       ¥1,045.00
    规格:
    More
    Specifications
    Cat. No.
    VWRI113-1143
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory
  • 14
    白色
    5XL
    Log in 查看库存
    Log in 查看库存
    1 * 25 件
    1 * 25 件 - ¥1,955.00
    ¥1,955.00
    1 * 25 件
       ¥1,955.00
    规格:
    More
    Specifications
    Cat. No.
    VWRU414004-703
    关闭类型
    带按扣紧固
    带口袋
    Garment application
    Cleanroom|Laboratory