洁净室防护罩袍,Basic,SPP
VWR® Basic SPP防护罩袍采用结实、轻质、透气的聚丙烯纺粘法(SPP)织物制成,能够为非危险环境提供防尘、防垢和特定干颗粒的防护。该防护罩袍适用于低临界区或预更衣进入间.
- 按扣领,插肩袖,按扣封口
- 松紧袖口,无口袋
- 轻质、透气的第一道阻隔防护
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
VWR® Basic SPP防护罩袍采用结实、轻质、透气的聚丙烯纺粘法(SPP)织物制成,能够为非危险环境提供防尘、防垢和特定干颗粒的防护。该防护罩袍适用于低临界区或预更衣进入间.
- 按扣领,插肩袖,按扣封口
- 松紧袖口,无口袋
- 轻质、透气的第一道阻隔防护
This garment is ideal for less critical areas or pre-gowning entry rooms. It is tested and manufactured in an ISO certified facility that is lot controlled and validated through independent laboratory testing.
- Catalog No:
- VWRI113-1152
- VWRI113-1151
- VWRI113-1154
- VWRI113-1155
- VWRI113-1153
- 关闭类型:
- 带按扣紧固
- 带按扣紧固
- 带按扣紧固
- 带按扣紧固
- 带按扣紧固
- 袋:
- 无口袋
- 无口袋
- 无口袋
- 无口袋
- 无口袋
- Gender:
- 中性
- 中性
- 中性
- 中性
- 中性
- Garment application:
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- Cleanroom|Laboratory
- 重量:
- 48 g/m²
- 48 g/m²
- 48 g/m²
- 48 g/m²
- 48 g/m²
Product Family Options
- 颜色衣服尺寸
Specifications
Cat. No.VWRI113-1152关闭类型带按扣紧固袋无口袋Gender中性Garment applicationCleanroom|Laboratory重量48 g/m²Specifications
Cat. No.VWRI113-1151关闭类型带按扣紧固袋无口袋Gender中性Garment applicationCleanroom|Laboratory重量48 g/m²Specifications
Cat. No.VWRI113-1154关闭类型带按扣紧固袋无口袋Gender中性Garment applicationCleanroom|Laboratory重量48 g/m²Specifications
Cat. No.VWRI113-1155关闭类型带按扣紧固袋无口袋Gender中性Garment applicationCleanroom|Laboratory重量48 g/m²Specifications
Cat. No.VWRI113-1153关闭类型带按扣紧固袋无口袋Gender中性Garment applicationCleanroom|Laboratory重量48 g/m²