Orders LinkContactUsLinkComponent
Small scale adapter (SSA)
12, Spindle SC4-18, 1,32 min⁻¹ min-1
  BROOSC4-18
Marketsource
 :   AMETEK
Small scale adapter (SSA)
  BROOSC4-18
 :  AMETEK
Marketsource
Small scale adapter (SSA)
Small scale adapter (SSA)
12, Spindle SC4-18, 1,32 min⁻¹ min-1
1 * 1 件
The SSA consists of a small cylindrical sample chamber and spindle with defined geometries for accurate measurement.
  • Water jacket for a rapid and precise control of temperature
  • Embedded temperature probe in sample chamber
  • Disposable sample chambers for difficult to clean samples
Delivery information: Supplied with water jacket, mounting bracket, sample chamber, and storage case.
  • 回转:
    1,32 min⁻¹ min-1
  • 说明:
    Spindle SC4-18
  • 顺序:
    12