Orders LinkContactUsLinkComponent
Immersion sampler
Immersion sampler
02, Immersion cylinder, 4300 g, 镀镍黄铜, 1000 ml, 450 mm, 75 mm
目录编号 BURK5365-7000
供应商:   BURKLE
Immersion sampler
目录编号 BURK5365-7000
供应商:  BURKLE
Immersion sampler
Immersion sampler
02, Immersion cylinder, 4300 g, 镀镍黄铜, 1000 ml, 450 mm, 75 mm
Price
询价
Log in 查看库存
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Completely made from non spark-producing materials, nickel plated.
  • Screw-off top enables easy cleaning
  • High weight for fast lowering
  • Robust handle
Immersion cylinder for sampling crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum, and liquid lubricants in explosion categories IIA, IIB, and IIC.
Point sampling: Two lowering cables are required for this (bearer cable and hauling cable). Lower the immersion cylinder to the required depth. When the target has been reached, pull on the hauling cable, the valve opens. The sample flows into the bottle. Release the hauling cable. The valve closes. Pull the cylinder back on the bearer cable. Empty the cylinder via the valve at the bottom.
Bottom sampling: Lower the immersion cylinder on the cable to the ground. The valve opens automatically because of the immersion cylinder's own weight. The sample flows into the cylinder. Pull the cylinder back on the cable, the valve closes independently. Empty the cylinder via the valve at the bottom.
Certifications: Tested for use in the Ex protection zone 0.
Ordering information: Please order the appropriate lowering cable at the same time.
规格
  • 直径:
    75 mm
  • 容量:
    1000 ml
  • 说明:
    Immersion cylinder, 4300 g
  • 长度:
    450 mm
  • 材质:
    镀镍黄铜
  • 顺序:
    02
Accessories
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0150
    12
    Stainless steel lowering cable with PTFE coating, Ø = 2 mm
    Log in 查看库存
    1 * 50 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0202
    08
    PP lowering cable, Ø = 2 mm
    Log in 查看库存
    1 * 25 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0510
    15
    Steel lowering chain, Ø = 2,3 mm
    Log in 查看库存
    1 * 10 米
产品信息
  • 目录编号
    顺序
    说明
    长度
    可用库存
    Price
  • BURK5309-0450
    05
    Copper lowering cable, Ø = 4,5 mm
    50 m
    Log in 查看库存
    1 * 50 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0525
    16
    Steel lowering chain, Ø = 2,3 mm
    Log in 查看库存
    1 * 25 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0550
    17
    Steel lowering chain, Ø = 2,3 mm
    Log in 查看库存
    1 * 50 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0125
    11
    Stainless steel lowering cable with PTFE coating, Ø = 2 mm
    Log in 查看库存
    1 * 25 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0110
    10
    Stainless steel lowering cable with PTFE coating, Ø = 2 mm
    Log in 查看库存
    1 * 10 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK0006-5301
    01
    Brass shackle
    Log in 查看库存
    1 * 1 件
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0425
    04
    Copper lowering cable, Ø = 4,5 mm
    Log in 查看库存
    1 * 25 米
产品信息
  • 目录编号
    顺序
    说明
    长度
    可用库存
    Price
  • BURK5309-0305
    02
    Stainless steel lowering cable, Ø = 1,25 mm
    50 m
    Log in 查看库存
    1 * 50 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0205
    09
    PP lowering cable, Ø = 2 mm
    Log in 查看库存
    1 * 50 米
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK0006-5300
    14
    Stainless steel snap hook
    Log in 查看库存
    1 * 1 件
产品信息
  • 目录编号
    顺序
    说明
    可用库存
    Price
  • BURK5309-0302
    13
    Stainless steel lowering cable, Ø = 1,25 mm
    Log in 查看库存
    1 * 25 米