Handheld refactometers
30GS, 0 - 85 °Bx ; 1,32 - 1,62 nD, 0,1 °Bx ; ±0,0005 nD, 0,0001 nD
目录编号 METT51324660
供应商: METTLER TOLEDO
Jump to:
Product Details & Documents
These refractometers, which use the total reflection method, are handheld instruments for measurements in the laboratory and production environments. The backlit LC display and the simple intuitive menu structure allow the user to easily change settings in seconds. The sample can either be placed on the measurement cell or the sensor can be immersed directly into the fluid.
- Selectable automatic temperature compensation (ATC)
- Handheld or benchtop instrument
- Choice of use: Immerse sensor in liquid or add sample to cell
- Storage of up to 1100 results and easy transfer with IR interface (adapter accessory 635-0117 required)
- Refracto GS has sapphire prism and thus extended range and faster response
The results can be displayed in a wide variety of standard units and also units applicable to antifreeze and wine measurement as well as customised applications.
Delivery information: Supplied with carry case, HelloCD™ software, two AAA batteries, cleaning tissues, two LDPE pipettes, two PE vials and operating instructions.
规格
- 型号:30GS
- 范围:0 - 85 °Bx ; 1,32 - 1,62 nD
- 分辨率:0,0001 nD
- 精度:0,1 °Bx ; ±0,0005 nD
- 重量:250 g
- Cat. No.:METT51324660
- 电气:Two AAA batteries
- 接口:Infrared for printer or PC
- 操作温度:10...40 °C
- 响应时间:3 - 180 seconds