Single-channel pipettes, electronic, variable volume, Finnpipette™ Novus
LABS46200300
- 颜色代码:黄色
- 容积:5 - 50 µl
- 顺序:04
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:ISO 14001 (Certificate Registration No. EMS 623400)
- Increment:0,1 µl
- 准确度:±3,0 - ±0,8 %
- 不准确度:2,5 - 0,3 %
- 包装规格:1
- 用于 Finntip™ 尖端:Flex 200, 250 Univ., 200 Ext.
This electronic pipette offers improved ergonomics and performance for standard pipetting applications.
- Light, ergonomic design for ease of use
- Finger rest can be pivoted through 120° so that pipetting can be controlled with the index finger
- Soft-touch eject button suitable for right- and left handed use
- Large screen, two level menu can be accessed with a single button
- Ten pipetting functions and nine speed levels can be accessed via a single button
- Autoclavable (end fitting only)
: Supplied with a universal AC adapter, a complete operating and maintenance manual, a certificate of calibration (according to ISO 8655) and a shelf hanger.