Multi-channel pipettes, electronic, variable volume, Finnpipette™ Novus
LABS46300800
- 颜色代码:绿松石
- 容积:100 - 1200 µl
- 顺序:04
- Environmentally Preferable:Low Manufacturing Impact
- 环境认证:ISO 14001 (Certificate Registration No. EMS 623400)
- Increment:1 µl
- 准确度:±3,0 - ±1,0 %
- 不准确度:0,9 - 0,2 %
- 包装规格:1
- 用于 Finntip™ 尖端:Flex 1200 µl
Ergonomic design for exceptionally easy use in routine laboratory work.
- Handle can be pivoted through 120° so that pipetting can be controlled with the index finger
- Soft-touch eject button suitable for right- or left handed use
- Large screen, two level menu can be accessed with a single button
- Ten pipetting functions and nine speed levels accessible via a single button, enables fast programming
- Autoclavable (end fitting only)
: Supplied with a universal AC adapter, a complete operating and maintenance manual, a certificate of calibration (according to ISO 8655) and an adhesive mini-support.