ContactUsLinkComponent
Multi-channel pipettes, mechanical, variable volume, mLINE®
Multi-channel pipettes, mechanical, variable volume, mLINE®
Catalog # BHIT725120
Supplier:  SARTORIUS BIOHIT
   Low Manufacturing Impact
undefined
Multi-channel pipettes, mechanical, variable volume, mLINE®
Catalog # BHIT725120
Supplier:  SARTORIUS BIOHIT
   Low Manufacturing Impact
Specifications
  • 容积:
    1 - 10 µl
  • 顺序:
    01
  • Environmentally Preferable:
    Low Manufacturing Impact
  • 环境认证:
    Renewable energy certificate; ISO 14001
  • Increment:
    0,01 µl
  • 准确度:
    ±1,50 %
  • 不准确度:
    <1,00 %
  • 包装规格:
    1
  • 用于吸液管尖端:
    10 / F10 µl
  • 带安全圆锥过滤器:
    -
Specifications
About this item
mLINE® pipettes offer a unique combination of new features for higher accuracy and precision. They include built-in replaceable tip-cone filters, which prevent pipette contamination and damage.
  • Clear and easy to read display with big numbers and small increments
  • Equipped with special spring-loaded tip cones (Optiload mechanism) allowing tip loading with equal constant force to every channel using minimum force
  • Plunger technology results in the lowest pipetting force
  • Volume range colour coding for easy pipette selection, and five colour coded caps
  • Thermal insulation of the internal components improves both accuracy and reproducibility
  • Autoclavable
Two types of filters are offered: Safe-Cone Standard filter and Safe-Cone Plus filter. It is recommended to use the Standard filter for general applications and the Plus filter for more demanding applications, such as cell culture, bacterial, viral work and molecular biology.
* Safe-Cone Plus